(4-Methoxyphenyl)acetic acid for synthesis, Sigma-Aldrich®

Danger

Synonyms: 4-Methoxyphenylacetic acid

M-Clarity™ quality level = MQ200

8.21240.0100 8.21240.0100 8.21240.0250 8.21240.0250
EM8.21240.0100CS 1602.03 USD
EM8.21240.0100 EM8.21240.0250 CA8.21240.0100 CA8.21240.0250
(4-Methoxyphenyl)acetic acid for synthesis, Sigma-Aldrich®
(4-Methoxyphenyl)acetic acid
Formula: (C₆H₄(OCH₃))CH₂COOH
MW: 166.18 g/mol
Boiling Pt: 306 °C (1013 hPa)
Melting Pt: 84…86 °C
Flash Pt: 193 °C
Storage Temperature: Ambient
MDL Number: MFCD00004345
CAS Number: 104-01-8
EINECS: 203-166-4

Order Now

Specification Test Results

Solubility 6 g/l (20 °C)
Melting Point 86 - 88 °C
Molar Mass 166.18 g/mol
Bulk Density 440 kg/m3
Boiling Point 138 - 140 °C (4 hPa)
Flash Point 193 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR