UltraClass Gold Alloy 8 Precision Analytical Weight Sets, Troemner
7234-UCGT 7236-UCG 7229-UCG 7221-UCGT 7227-UCGW 7226-UCGW 7222-UCGT 7228-UCGT 7222-UCGW 7231-UCGT 7221-UCG 7225-UCG 7224-UCGT 7240-UCG 7220-UCG 7218-UCGT 7237-UCGW 7218-UCG 7235-UCGW 7233-UCG 7238-UCGT 7232-UCGW 7219-UCGT 7224-UCGW 7217-UCGW 7236-UCGW 7231-UCGW 7232-UCG 7227-UCGT 7240-UCGT 7228-UCGW 7239-UCGW 7220-UCGT 7235-UCGT 7238-UCGW 7233-UCGT 7219-UCG 7234-UCGW 7236-UCGT 7228-UCG 7222-UCG 7225-UCGW 7217-UCG 7220-UCGW 7229-UCGT 7232-UCGT 7234-UCG 7237-UCG 7225-UCGT 7224-UCG 7221-UCGW 7218-UCGW 7235-UCG 7230-UCGW 7231-UCG 7226-UCGT 7230-UCG 7229-UCGW 7240-UCGW 7230-UCGT 7237-UCGT 7219-UCGW 7227-UCG 7238-UCG 7239-UCG 7239-UCGT 7226-UCG 7217-UCGT 7233-UCGW 7223-UCGW 7223-UCG 7223-UCGT
10026-368EA 2186.53 USD
10026-368 10026-764 10026-642 10026-686 10026-400 10026-766 10026-644 10026-688 10026-402 10026-364 10026-682 10026-762 10026-366 10026-684 10026-640 10026-680 10026-638 10026-654 10026-378 10026-334 10026-774 10026-656 10026-336 10026-776 10026-650 10026-694 10026-770 10026-374 10026-330 10026-652 10026-410 10026-696 10026-772 10026-332 10026-376 10026-690 10026-370 10026-692 10026-372 10026-408 10026-328 10026-768 10026-646 10026-404 10026-406 10026-648 10026-346 10026-664 10026-666 10026-342 10026-386 10026-660 10026-388 10026-344 10026-662 10026-382 10026-384 10026-340 10026-380 10026-658 10026-338 10026-676 10026-678 10026-672 10026-396 10026-674 10026-398 10026-392 10026-670 10026-394 10026-390 10026-668
UltraClass Gold Alloy 8 Precision Analytical Weight Sets, Troemner
Highly polished kilogram and gram weights made of Troemner Alloy 8, a specially developed austenitic stainless steel that possesses closely controlled density, extremely low magnetic permeability, good stability, and resistance to corrosion and handling damage
Fractional weights 500 mg to 5 mg are made of stainless steel (30 mg weights are made of aluminum); fractional weights 3 mg and below are made of aluminum. Analytical weights 1 g and above are manufactured with adjusting cavities. Weights 500 g and smaller are supplied in rugged polypropylene cases. A clear fitted cover secures the fractional weights within the case. Sets that include weights 1 kg and larger are supplied in oak cases. All cases are fitted with individual compartments and marked with the accuracy class and serial number. Each set includes contoured weight forceps.
This exclusive material has a density of 8.0 at 25°C (controlled to ±0.1%), is essentially nonmagnetic in the annealed condition, and will not transform to magnetic under any combination of temperature and cold work down to –195.5°C (–320°F). UltraClass Gold weight tolerances are equal to ANSI/ASTM E617 Class 00 and exceed OIML R 111 Class E1 tolerances. UltraClass Gold weights combine high precision with the advantage of two-piece construction avoiding costly replacement issues associated with one-piece weights.
Información para pedidos: Standard weight sets are supplied with a Statement of Accuracy. Where applicable, weight sets are supplied with a NIST/NVLAP Weight Calibration Certificate or a Traceable Certificate.