Prime Full Size Thermal Cyclers, Techne®
93945-15 93945-71 93945-13 93945-23 93945-21 93945-17 93945-19 93945-73
10066-590EA 5605.49 USD
10066-590 89235-830 10066-592 89235-812 89235-826 89235-816 89235-814 89235-828
Prime Full Size Thermal Cyclers, Techne®
Thermal Cyclers
The Prime full size thermal cycler delivers high performance, high throughput and maximum flexibility with an ability to upgrade base model to gradient capability with a simple unlock code, as well as the availability of a wide choice of thermal blocks.
User friendly programming is achieved via a colour touchscreen and intuitive software that is standardised across the entire Techne® thermal cycler range. A USB port enables the transfer of programmes between instruments and temperature logs to your PC. A real time graphical display provides an instant visualisation of the program status.
Optional gradient cycling can be achieved directly using the PrimeG instrument or by upgrading the Prime unit via a USB software installation. This enables the laboratory to manage changing cycling requirements in a cost effective manner. The Techne® Prime cycler range is one of the most affordable full sized thermal cyclers on the market and is backed up by a 4 year warranty.
Información para pedidos: Can be gradient enabled with USB upgrade.