Radiometer Analytical PHC2085-6 Combination Red-Rod pH Electrode with Temperature Sensor, Epoxy Body, Hach
E16M345
89174-252EA 845.81 USD
89174-252
Radiometer Analytical PHC2085-6 Combination Red-Rod pH Electrode with Temperature Sensor, Epoxy Body, Hach
The Radiometer PHC2085-6 Combination pH Electrode has a robust epoxy body, Red-Rod reference system, refillable electrolyte (sat
  • The right probe for your application.
  • Unique RedRod technology provides a reproducible, fast and accurate response.
  • Know that you have the right answer.
  • Avoid problems with your measurement, even in challenging samples types.
  • Measure over a wide temperature range.
The PHC2085-6 has a fixed cable with BNC + RJ9 connectors for use with Hach sensION+ benchtop pH meters.
KCl), integrated temperature sensor, and porous pin junction for general purpose applications.
Información para pedidos: Radiometer Analytical PHC2085-6 combination pH electrode with Certificate of Conformity and small bottle of internal electrolyte solution included.