SP Bel-Art Dry-Keeper™ Vertical Desiccator Cabinet, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a part of SP
H42056-0001
24983-397EA 1696.89 USD
24983-397
SP Bel-Art Dry-Keeper™ Vertical Desiccator Cabinet, Bel-Art Products, a part of SP
Desiccators Desiccator Cabinets
Clear and durable PVC desiccator cabinet has a generous 2 cu
  • Clear PVC construction
  • Durable
  • Adjustable shelves
  • Dial hygrometer provides easy-to-read measurements
Three adjustable, removable, and perforated shelves facilitate full circulation of air and rest on shelf rails that can be repositioned on any one of 15 steps set at 28 mm intervals. Additional shelves and rails sold separately. A large, in-door dial hygrometer provides easy visualization of rH%. Gasketed door opens to the right and closes with two latches for a tight seal. The interior opening measures 25 x 43 cm (10 x 17").
ft. interior volume, sleek black ABS accents, and movable shelves for adjusting dry storage space configuration for moisture-sensitive materials.
Información para pedidos: Includes desiccant tray (desiccant sold separately, recommend F42048-0100).