Inlet/Thermometer Adapter, Glassco
Supplier: Glassco
056.467.01
89426-438EA 1.97 USD
89426-438
Inlet/Thermometer Adapter, Glassco
Adapters Adapters, Conical Ground Joints
Adapter is equipped with a compression cap and Viton O-Ring to form a vaccum-tight seal.
  • Forms a vacuum-tight seal
  • Compression cap and o-ring included
  • Accommodates thermometer
To use as a syringe port, simply replace the O-Ring with fluoropoymer resin-lined septum. Cap accommodates 5.0–7.5mm O.D.