Shipping Label with Generator Information 'Hazardous Waste', New Pig
89135-004
: New Pig
- Description:Shipping label with generator information
- Dimensions:152×152 mm (6×6")
- Material:Paper
- Cat. No.:89135-004
- Legend:Hazardous Waste
- Supplier no.:SGN659-PPR