Horizontal Gel Casters, Expedeon
GCT-14-162 GCT-20-301 GCT-14-201 GCT-10-112 GCT-20-202 GCT-10-111 GCT-20-401 GCT-20-302 GCT-26-261 GCT-14-101 GCT-26-401 GCT-26-141 GCT-20-201 GCT-20-402 GCT-14-161 GCT-26-262 GCT-14-102 GCT-26-142 GCT-26-402 GCT-14-202
CBGCT-26-142EA 148.6 USD
CBGCT-26-142 CBGCT-26-141 CBGCT-26-262 CBGCT-26-261 CBGCT-14-161 CBGCT-14-162 CBGCT-10-112 CBGCT-26-402 CBGCT-26-401 CBGCT-20-401 CBGCT-20-302 CBGCT-14-202 CBGCT-20-402 CBGCT-10-111 CBGCT-14-101 CBGCT-20-201 CBGCT-20-301 CBGCT-14-201 CBGCT-20-202 CBGCT-14-102
Horizontal Gel Casters, Expedeon
External gel casters allow fast easy casting outside the horizontal gel unit
These support trays have leveling bases and are lined with soft, coated neoprene to provide firm yet supple support for casting gels without leakage. 1 or 2 place casters are available for every size horizontal unit.