Model EN300 Hygro-Thermo-Anemometer-Light-Sound Meter, Extech
Supplier: Extech
EN300
89370-180EA 654.58 USD
89370-180
Model EN300 Hygro-Thermo-Anemometer-Light-Sound Meter, Extech
Anemometers
The Hygro-Thermo-Anemometer-Light-Sound Meter measures humidity, temperature, air velocity, light, and sound
Model EN300 features data hold, min./max., auto power off, and low battery/overrange indication functions. The large, dual LCD simultaneous displays temperature and air velocity or relative humidity. Built-in low friction vane wheel improves the accuracy of air velocity in ft./min., mph, m/s, km/h, and knots. An integrated precision thin-film capacitance humidity sensor enables fast responses. Unit also features thermistor for ambient temperature measurements.
Certificaciones: CE marked.
Información para pedidos: Supplied complete with built-in sensors, Vane wheel, windscreen, Type K thermocouple probe, pouch case, and six 1.5V AAA batteries. Unit also includes manufacturer's one-year warranty.