Pocket Colorimeter™ II, Wavelength Specific Model, 580 nm, Hach
5870058
97026-962EA 616.3 USD
97026-962
Pocket Colorimeter™ II, Wavelength Specific Model, 580 nm, Hach
Generic Pocket Colorimeter II Instruments are intended for applications not included in standard analysis systems
  • Single Parameter go anywhere portable photometer
  • USEPA approved for drinking water and wastewater
  • Factory-Programmed
  • Waterproof, Lightweight, Rugged
  • Backlit Display, Data Logging, Recall
  • Two year manufacturer's warranty
They are available in a range of wavelengths appropriate for many custom calibrations. In addition to the factory absorbance calibration (0-2.5 A), the instruments offer two user-entered, two-point, linear calibrations, and provide a choice of either 0.00 to 2.55 or 0.0 to 99.0 for the display value.
Información para pedidos: 2 yr warranty.