Differential Pressure Manometer, Heavy Duty, Extech, Flir
Supplier: Flir
407910
89370-218EA 511.53 USD
89370-218
Differential Pressure Manometer, Heavy Duty, Extech, Flir
Manometers
Wide-range (0 to 29psi) Gauge/Differential Pressure Meter with 8 Selectable Units of Measure
Features a zero function for offset measurements. Other features include Data Hold, Auto power off, and Min/Max functions. Also comes with its own Built-in RS-232 PC interface.
Certificaciones: CE marked.
Información para pedidos: Three year warranty. Complete with 2 quick disconnect-fittings, 9V battery, protective rubber holster with stand, and carrying case.