Would you like to log in again?
BART Test for Denitrifying Bacteria, Hach
Supplier: Hach
2619309
97026-618PK 166.8 USD InStock
97026-618
BART Test for Denitrifying Bacteria, Hach
Microbiology Test Systems Microorganism Tests
BART biodetectors are excellent diagnostic tools to help identify the presence and activity of various bacteria.
  • Simple yet effective method for monitoring the population size and/or activity of specific groups of bacteria
  • Easy to use, requiring no elaborate or costly equipment and no specialized training
  • Effective and affordable tests are easy to interpret and can be performed at room temperature in virtually any environment
Denitrifying bacteria indicate the decomposition of waste organic nitrogenous materials. These bacteria reduce nitrate to nitrite and some continue nitrification to gaseous nitrogen (complete denitrification). In water, aggressive denitrifiers can indicate high concentrations of nitrates, and that the sample is probably anaerobic and relatively rich in organic matter. The presence of denitrifying bacteria can indicate that the water has been polluted by nitrogen-rich organics from sources such as compromised septic tanks, sewage systems, industrial and hazardous waste sites. If highly aggressive bacteria are detected, the water should be tested for the presence of coliform bacteria.
Información para pedidos: 9/pk.
Order Now
ORDER
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty