PFA Tongs, Savillex Corporation
Supplier: Savillex
700-850
30617-546EA 618.17 USD
30617-546
PFA Tongs, Savillex Corporation
Tongs
Adjustable PFA tongs are molded from the highest purity PFA resin for ultratrace metals use
Service temperature range is –200 to 260°C (–328 to 500°F).
Ideal for transferring ICP glassware during cleaning. Tongs are 28cm (11") long with three adjustable width positions.