Pocket Digital Refractometer, Custom 1 Scale, Sper Scientific
300056
97011-848EA 581.83 USD
97011-848
Pocket Digital Refractometer, Custom 1 Scale, Sper Scientific
Select from 94 custom scales to measure concentrations in brix, salinity, and a wide variety of other parameters in the field with laboratory accuracy
As small, light, and simple to use as optical refractometers, but with the added precision of a digital readout. Reads samples as small as 1mL. Sample temperature is initially displayed on the large LCD followed by the measurement results. The sophisticated microprocessor in this digital refractometer provides automatic calibration, automatic temperature compensation, and automatic shutoff (after 3 minutes). A sturdy water-resistant case protects the electronics from wet environments and makes for easy clean ups. All models feature one button calibration to distilled water and indicate when the battery should be replaced (8,000 readings).
Información para pedidos: Supplied ready to use with distilled water, transfer-pipet, AAA battery, instructions, and a carrying case. The refractometer also includes a 1-year warranty. Please specify desired scales when ordering.