The Visible Light SD Card Logger reads lux, foot candles, and functions as a type K or J thermometer
The unit records data onto standard SD memory cards (like a digital camera). When full, simply exchange SD cards for unlimited data storage. The removable SD card is easy to transport and insert into a computer’s memory card slot or SD card adapter. It uploads preformatted data to Excel, without the need for additional software and cables. Additional SD cards are readily available in a variety of memory sizes. The card logger can also stream data directly to a computer using the optional software and cables. Each data set includes lux or foot candle reading, time, and date. It features push button zero adjustment, min./max., auto power off, low battery indicator, and hold function. This well-made unit is extremely accurate with fast response times, and includes a fold-out tripod and large, easy-to-read back-lit LCD.
Información para pedidos: Supplied ready to use in a soft carrying case, complete with a detachable color-corrected probe that meets the C.I.E. photopic spectrum, 6 AA batteries, instruction manual, and a 2GB SD card. Each unit includes a 5-year warranty.