JACKSON SAFETY® AA-9 WINDGARD™ Head Protection, KIMBERLY-CLARK PROFESSIONAL®
16757 14497
33001-152CS 993.83 USD
33001-152 33001-154
JACKSON SAFETY® AA-9 WINDGARD™ Head Protection, KIMBERLY-CLARK PROFESSIONAL®
Keeps wind out and traps warmth
  • For temperatures ranging from 0 to –20° F (–20 to –28°C)
  • Knitted, flame-resistant, treated polyester meets 16 CFR 1610, Class 1 requirements
  • High-visibility color
  • Full face and neck model protects mouth and cheeks
  • Fits over most safety caps
Knitted, flame-resistant, treated polyester meets 16 CFR 1610, Class 1 requirements. High-visibility color. Full face and neck model protects mouth and cheeks. Fits over most safety caps. Warmth for temperatures ranging from 0 to –20° F (–20 to –28°C).