Would you like to log in again?
Hand-Held Series Portable Balance, Ohaus®
Supplier: OHAUS SCALE CORP
71142841 71142844 HH120
14005-802EA 220.02 USD InStock
14005-802 14005-800 11377-234
Hand-Held Series Portable Balance, Ohaus®
Balances and Scales
Convenient, compact and portable weighing capability designed to fit in the palm of your hand
Standard balance features include, low battery indicator, overload and underload indicators, protective snap-on cover, external push button calibration, and button-activated luminescent display on HH120 and HH320 models. Hand-Held balances are constructed of ABS housing, with an ABS protective cover and ABS pan.
These little scales with sleek styling and convenient sizes are ideal to take anywhere. They are compact enought to be thrown in a briefcase or dropped in a backpack. Whether you choose the HH120D model with basic grams weighing, or the HH120 and HH320 models with multiple weighing units, the Ohaus® Hand-Held series ensures outstanding performance and exceptional value.
Información para pedidos: Balances are supplied with 2 AAA batteries and a protective snap-on cover.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty