Quantabio Genomics Products
Quantabio is a leading provider of advanced DNA and RNA amplification reagents for the most demanding molecular testing applications in applied, translational, and life science research. Following a partnership with Invitrogen and as a formaer manufacturer for BioRad Laboratories, Quantabio leverages decades of scientific experience and a legacy of innovation to develop cutting-edge amplification enzymes and reagents, such as SuperScript® I/II and Platinum® Taq. 
Fast forward to 2018: Quantabio, based in Beverly, Mass., offers a growing portfolio of products through its exclusive partnership with VWR International to continue bringing innovations to market. Quantabio continues building on its foundational qPCR and cDNA synthesis product brands (PerfeCTa®, ToughMix® and qScript®) with expansion into Next Generation Sequencing, Cloning, and qPCR instrumentation to help set new assay performance & cost standards in critical PCR, qPCR, and NGS-based applications.
Q qPCR Cycler
NEW Q qPCR cycler redefines runtime to as little as 25 minutes while enhancing performance for molecular testing applications.
sparQ Next Generation Sequencing (NGS) Solutions
sparQ NGS products simplify workflow steps while improving library yields with limiting sample inputs, resulting in lower total sequencing costs.
ToughMix® qPCR MasterMixes
ToughMixes neutralize problem-causing inhibitors present in crude samples, avoiding expensive and time-consuming purification steps.
qScript® cDNA Synthesis Kits
qScript cDNA synthesis kits use advanced reverse transcriptase technology for maximum yield and sensitivity for low-abundance RNA detection.