Fire Assay
All of your fire assay needs are available through VWR. Consumable items such as Alpine Cupels, APGreen and Anachemia crucibles, custom made flux, fluxing reagents including borax, soda ash, silica and litharge.
We offer the Mines Assay 3 electric fusion and cupellation furnace, as well as larger gas cupellation and fusion furnaces. We also offer a single head or dual head flux dispenser offering fast and safe delivery of your flux. Tumbler mixers are also available to ensure fast and efficient mixing of your sample with the flux.