Azure cSeries Advanced Imaging Systems
The c300-c600 offer imaging capabilities for chemiluminescence, UV, RGB, NIR, blue light, and white light, depending on the model that fits your needs. The c600 offers the most flexible imaging options, but each model can be upgraded to suit your lab’s needs as they change.
c600 c500 c400 c300
Ultimate Western System
IR Fluorescence
Visible Fluorescence
Chemiluminescence
UV Fluorescence
Visible Imaging
Blue Excited DNA Dyes
Infrared Imaging System
IR Fluorescence
Chemiluminescence
UV Fluorescence
Visible Imaging
Blue Excited DNA Dyes
Upgradeable to c600
Visible Fluorescent
Western System
Visible Fluorescence
Chemiluminescence
UV Fluorescence
Visible Imaging
Blue Excited DNA Dyes
Upgradeable to c600
Darkroom Eliminator
Chemiluminescence
UV Fluorescence
Visible Imaging
Blue Excited DNA Dyes
Upgradeable to c600, c500, and c400
cSeries Specifications c600 c500 c400 c300
Camera 8.3 MP
Cooling -50°C regulated cooling
7 Position Filter Wheel
UV 302/365 nm
EPI Blue LED
Chemi
Visible Fluorescent Westerns    
NIR Westerns    
Field of View 20 × 15 cm
Footprint (W × H × D) 38 × 55 × 36 cm
Product Selection