Hypodermic and customizable needles
Air-Tite Products provides one of the largest selections of hypodermic needles from multiple manufacturers for a wide variety of uses. Whether you are looking for medical, laboratory, or industrial applications, we strive to have the gauge and length you are seeking. Need a custom size? Contact your VWR rep to work with Air-Tite for a customized solution matching your exact needs.
Product selection
Air-Tite needles
14 gauge to 27 gauge Air-Tite brand needles for medical, laboratory, industrial, and veterinary use.
TSK needles
Premium-quality 31g, 32g, and 33g STERiJECT® Japanese needles from TSK.