VWR® Atlas Slot-Mounted Shelving, Single-Sided
Supplier: VWR, part of Avantor
VWR®
VWRSSNL1248 VWRSSNL1260 VWRSSNL1242 VWRSSNL1236 VWRSSNL1272 VWRSSNL1266 VWRSSNL1854 VWRSSNL1542 VWRSSNL1848 VWRSSNL1572 VWRSSNL1548 VWRSSNL1842 VWRSSNL1254 VWRSSNL1860 VWRSSNL1866 VWRSSNL1536 VWRSSNL1566 VWRSSNL1554 VWRSSNL1836
VWRSSNL1248EA 401.97 USD
VWRSSNL1248 VWRSSNL1260 VWRSSNL1242 VWRSSNL1236 VWRSSNL1272 VWRSSNL1266 VWRSSNL1854 VWRSSNL1542 VWRSSNL1848 VWRSSNL1572 VWRSSNL1548 VWRSSNL1842 VWRSSNL1254 VWRSSNL1860 VWRSSNL1866 VWRSSNL1536 VWRSSNL1566 VWRSSNL1554 VWRSSNL1836
VWR® Atlas Slot-Mounted Shelving, Single-Sided
Furniture Shelves
Slot-mounted shelving for single-sided applications.
  • Color: Light neutral
Slot-mounted shelving is used with slotted uprights (see VWRSU24 series) for shelf adjustment in 2.5 cm (1") increments.