VWR® Modular Workstations, 60W×30D×30H"
Supplier: VWR, part of Avantor
B603030F B603030L B603030E B603030EB B603030PB B603030P
VF-B603030EEA 2947.82 USD
VF-B603030E VF-B603030F VF-B603030EB VF-B603030PB VF-B603030P VF-B603030L
VWR® Modular Workstations, 60W×30D×30H"
Furniture Workbenches
VWR® modular work benches are designed for a wide range of applications in laboratory and industrial environments including biotech, pharmaceutical, analytical, material handling, distribution, and assembly.
  • Can be custom configured
  • Shipped fully assembled
  • Includes a variety of attachments
Standard work benches handle 1000lbs
The VWR® Basic Bench includes 4 standard leveler bench feet, gussets and light neutral powder coat of 16 gauge 1.5x1.5” steel frame. Shipped fully assembled. Also available in 48" and 72" widths.
of evenly distributed weight but can be configured to handle loads of 5000lbs. or more. Tables can be custom configured with numerous accessories including, 8" adjustable leg kits (for ADA compliance), locking casters, heavy-duty 2x2 steel frame, upper and lower shelves with pencil lips, backsplashes, lighting, bin rails, power strips, ESD tops, and steel drawers.
Información para pedidos: Select work bench size followed by type of work surface (epoxy, plastic laminate, ESD, phenolic). Next add appropriate accessories to the bench to complete your configuration. If you need something not on the price list, then please contact your VWR sales representative. Exact frame dimensions depend upon reveal and thickness of work surface to be added.