Quantus™ Fluorometer, Promega
Supplier: Promega
SA4040 E6150
PASA4040EA 717.66 USD
PASA4040 PAE6150
Quantus™ Fluorometer, Promega
Spectrophotometers and Spectrometers Fluorescence Spectrophotometers
The Quantus Fluorometer is a dual-channel fluorometer for your personal quantitation workflow. Designed to provide highly sensitive fluorescent detection when quantifying nucleic acids, the compact instrument is simple to operate.
  • Simple, sensitive DNA and RNA quantitation
  • Easy, intuitive controls
  • Ready to use with QuantiFluor(R) dyes
  • Recommended for next-generation sequencing
The Quantus Fluorometer is a dual-channel fluorometer for your personal quantitation workflow. Designed to provide highly sensitive fluorescent detection when quantifying nucleic acids, the compact instrument is simple to operate. The Quantus Fluorometer is optimized with preprogrammed settings for Promega QuantiFluor Dye Systems (QuantiFluor dsDNA, RNA and ssDNA Systems) to quantitate nucleic acids and offers the flexibility to create customized methods and quantitation settings for other fluorescent dyes. The Quantus Fluorometer is equipped with two fluorescence channels for nucleic acid and protein quantitation: 1) Blue fluorescence channel: Excitation 495nm shortpass (wavelengths up to 495nm), emission 510-580nm; 2) Red fluorescence channel: Excitation 640nm shortpass (wavelengths up to 640nm), emission 660-720nm.
Información para pedidos: To learn more about the Quantus Fluorometer, please contact your VWR representative.