Microcaps® TLC Spotting Capillary Tubes, Kimble Chase
Supplier: DWK Life Sciences
764500-0002 764520-0000 764500-0025 764500-0001 764500-0020 764500-0050 764500-0100 764500-0000 764500-0010 764500-0005
KT764500-0002PK 63.91 USD
KT764500-0002 KT764500-0025 KT764500-0000 KT764500-0001 KT764500-0100 KT764500-0020 KT764520-0000 KT764500-0010 KT764500-0050 KT764500-0005
Microcaps® TLC Spotting Capillary Tubes, Kimble Chase
Pipets Microcapillaries
Microcaps® precision bore glass capillary tubes are cut to predetermined lengths so that each capillary tube will hold a known fluid volume when filled.
  • Made of borosilicate glass
  • Volumetric tolerance is ±1%
  • Fluid dispensed by simple squeeze of the bulb
  • Ideal for spotting preadsorbent TLC plates
Información para pedidos: Supplied with one bulb assembly and one dispenser vial with 100 micropipets.