Fastprep-96™ Sample Preparation Instrument, MP Biomedicals
Supplier: MP Biomedicals
116010500 116005510
IC116010500EA 25704 USD
IC116010500 CAIC116010500
Fastprep-96™ Sample Preparation Instrument, MP Biomedicals
Homogenizers Benchtop Homogenizers
MP Biomedicals' FastPrep-96™ is a versatile high-speed homogenizer offering the ultimate in high-throughput sample preparation
FastPrep-96™ is used to homogenize biological samples. FastPrep-96 Instrument lyses thoroughly and quickly any tissues and cells and thus allows easy and reproducible isolation of stable RNA, active proteins and full-length genomic DNA.
The FastPrep-96™ Instrument features High Throughput Processing of up to 192 samples simultaneously in 2 x 96 deep well plates. Versatile Interchangeable adapters available for 2 x 96 deep well plates, 96 x 2 mL tubes, 48 x 4.5 mL tubes, 24 x 15 mL tubes, 8 x 50 mL tubes, and 1 x 250 mL flask. Reproducibly Consistent lysis results in every well due to True Linear Motion that eliminates the need to re-orient plates mid-cycle. Fast Processing Speed because of its high-power, 1800 Oscillations/min and 1.5-inch stroke. Preservation of temperature sensitive molecules with homogenization performed at room or cryogenic temperatures..
Suitable for all sample types; Lysing highly contagious samples; Lysis of yeast and fungi; Used for Isolation of PCR-Ready Genomic DNA from Fecal Samples; Used for Isolation of PCR-Ready Genomic DNA from Fungi and Bacteria Samples; Used for Isolation of PCR-Ready Genomic DNA from Soil Samples; DNA & RNA Purification; Lysis of bacteria; Lysis of soft tissues; Isolation of genomic DNA; Purification of DNA; Cell Lysis; Used to lyse cell wall of yeast; Break hard/brittle samples; Used to isolate DNA from virtually any source
Developed for resistant samples, the FastPrep-96™ instrument uses a high-speed linear motion to disrupt thoroughly any tissues and cells in just seconds through the simultaneous beating of specialized Lysing Matrix beads on the sample material. This high-performance system saves hours of work during the sample preparation stage and allows for the purification of high yields of intact DNA, RNA, proteins and other metabolites.
Información para pedidos: FastPrep-96 Instrument, Power cord, Standard Dual Plate Holder (2 x 96 well block cap) User Manual.