Ion Chromatography (IC) Anion and Cation Check Standards, VWR Chemicals BDH®
Supplier: VWR Chemicals BDH®
BDH0837-125ML BDH0832-125ML BDH0835-125ML BDH0833-125ML BDH0836-125ML BDH0834-125ML
BDH0836-125MLEA 222.81 USD
BDH0836-125ML BDH0833-125ML BDH0837-125ML BDH0835-125ML BDH0832-125ML BDH0834-125ML
Ion Chromatography (IC) Anion and Cation Check Standards, VWR Chemicals BDH®
Standards Anionic Standards Multi-Element Anioinic Standards
Ion chromatography (IC) is an analytical process for the separation of ions based on charge affinity. IC can be used for a variety of different kinds of charged analytes from single elements to large proteins.
  • Multi-Element Standards for Ion Chromatography