Ammonium (NH4+) Single-Element Ion Cation Standard, 1,000 µg/ml (1,000 ppm), VWR Chemicals BDH®
Supplier: VWR Chemicals BDH®
BDH0831-125ML
BDH0831-125MLEA 73.12 USD
BDH0831-125ML
Ammonium (NH4+) Single-Element Ion Cation Standard, 1,000 µg/ml (1,000 ppm), VWR Chemicals BDH®
Standards Cationic Standards Ammonium Standards
Ion chromatography & ion selective electrode (ISE) standard.
  • Single and Multi-Element Standards for Ion Chromatography available
  • Stable and traceable