Phosphate-Phosphorus Single-Element Ion Anion Standard, 1,000 µg/ml (1,000 ppm), VWR Chemicals BDH®
Supplier: VWR Chemicals BDH®
BDH0824-125ML BDH0824-500ML
BDH0824-500MLEA 143.9 USD
BDH0824-500ML BDH0824-125ML
Phosphate-Phosphorus Single-Element Ion Anion Standard, 1,000 µg/ml (1,000 ppm), VWR Chemicals BDH®
Standards Anionic Standards Phosphate(P) Standards
Ion chromatography & ion selective electrode (ISE) standard.
  • Single and Multi-Element Standards for Ion Chromatography available
  • Stable and traceable