Phosphate (PO₄3-) Single-Element Ion Anion Standard, 1,000 µg/ml (1,000 ppm), VWR Chemicals BDH®
Supplier: VWR Chemicals BDH®
BDH0823-500ML BDH0823-125ML
BDH0823-125MLEA 73.12 USD
BDH0823-125ML BDH0823-500ML
Phosphate (PO₄3-) Single-Element Ion Anion Standard, 1,000 µg/ml (1,000 ppm), VWR Chemicals BDH®
Standards Anionic Standards Phosphate Standards
Ion chromatography & ion selective electrode (ISE) standard.
  • Single and Multi-Element Standards for Ion Chromatography available
  • Stable and traceable