Nitrate-Nitrogen Single-Element Ion Anion Standard, 1,000 µg/ml (1,000 ppm), VWR Chemicals BDH®
Supplier: VWR Chemicals BDH®
BDH0822-125ML BDH0822-500ML
BDH0822-500MLEA 143.9 USD
BDH0822-500ML BDH0822-125ML
Nitrate-Nitrogen Single-Element Ion Anion Standard, 1,000 µg/ml (1,000 ppm), VWR Chemicals BDH®
Standards Anionic Standards Nitrate(N) Standards
Ion chromatography & ion selective electrode (ISE) standard.
  • Single and Multi-Element Standards for Ion Chromatography available
  • Stable and traceable