Chlorate (ClO₃)- Single-Element Ion Anion Standard, 1,000 µg/ml (1,000 ppm), VWR Chemicals BDH®
Supplier: VWR Chemicals BDH®
BDH0815-500ML BDH0815-125ML
BDH0815-500MLEA 201.6 USD
BDH0815-500ML BDH0815-125ML
Chlorate (ClO₃)- Single-Element Ion Anion Standard, 1,000 µg/ml (1,000 ppm), VWR Chemicals BDH®
Standards Anionic Standards Chlorate Standards
Ion chromatography & ion selective electrode (ISE) standard.
  • Single and Multi-Element Standards for Ion Chromatography available
  • Stable and traceable