Bromide (Br-) Single-Element Ion Anion Standard, 1,000 µg/ml (1,000 ppm), VWR Chemicals BDH®
Supplier: VWR Chemicals BDH®
BDH0811-125ML BDH0811-500ML
BDH0811-125MLEA 73.12 USD
BDH0811-125ML BDH0811-500ML
Bromide (Br-) Single-Element Ion Anion Standard, 1,000 µg/ml (1,000 ppm), VWR Chemicals BDH®
Standards Anionic Standards Bromide Standards
Ion chromatography & ion selective electrode (ISE) standard.
  • Single and Multi-Element Standards for Ion Chromatography available
  • Stable and traceable