Toxaphene Standard for US EPA Methods 525, 625 and 8270
Supplier: VWR Chemicals BDH®
BDH0806-1ML
BDH0806-1MLEA 98.9 USD
BDH0806-1ML
Toxaphene Standard for US EPA Methods 525, 625 and 8270
Standards Environmental Analysis Standards
Semivolatiles standard.
US EPA Method 525 is an analytical method for the monitoring of semivolatiles in drinking water and raw water sources by GC/MS.
US EPA Method 625 is an analytical method for the monitoring of acid and base neutral extractable fractions in municipal and industrial wastewater by GC/MS.
US EPA Method 8270 is a general analytical method for the monitoring of semivolatiles in environmental and waste samples by GC/MS.