Trihalomethanes Volatiles Mix for US EPA Method 502.2
Supplier: VWR Chemicals BDH®
BDH0804-1ML
BDH0804-1MLEA 31.58 USD
BDH0804-1ML
Trihalomethanes Volatiles Mix for US EPA Method 502.2
Standards Environmental Analysis Standards
Volatiles standard.
US EPA Method 502.2 is a general analytical method for the monitoring of halogenated, purgeable VOCs in drinking water and raw water sources by GC megabore capillary column techniques and Hall and Photoionization Detectors (PID).