TCLP Volatiles Spike Standard for TCLP Series Methods
Supplier: VWR Chemicals BDH®
BDH0803-1ML
BDH0803-1MLEA 101.54 USD
BDH0803-1ML
TCLP Volatiles Spike Standard for TCLP Series Methods
Standards Environmental Analysis Standards
TCLP Series Volatiles standards.
Toxicity Characteristic Leaching Procedure (TCLP) is a soil sample extraction method for chemical analysis employed as an analytical method to simulate leaching through a landfill.