TCLP Base/Neutral/Acid Extractable Spike Solution for US EPA Method 1311-TCLP Series
Supplier: VWR Chemicals BDH®
BDH0802-1ML
BDH0802-1MLEA 72.98 USD
BDH0802-1ML
TCLP Base/Neutral/Acid Extractable Spike Solution for US EPA Method 1311-TCLP Series
Standards Environmental Analysis Standards
Semivolatiles standard.
US EPA Method 1311-TCLP Series Methods - EPA methods 1311 and 1312 analytical methods for the monitoring of a wide range of compounds in various samples using the Toxicity Characteristic Leaching Procedure (TCLP).