Aroclor 1248 for US EPA Methods 505, 508, 608, 8082, CLP Series Methods and ASTM Method D6160
Supplier: VWR Chemicals BDH®
BDH0790-1ML
BDH0790-1MLEA 54.56 USD
BDH0790-1ML
Aroclor 1248 for US EPA Methods 505, 508, 608, 8082, CLP Series Methods and ASTM Method D6160
Standards Organic Standards
Semivolatiles standard.
US EPA Method 625 is an analytical method for the monitoring of acid and base neutral extractable fractions in municipal and industrial wastewater by GC/MS.
US EPA Method 8270 is a general analytical method for the monitoring of semivolatiles in environmental and waste samples by GC/MS.