Acid Surrogates Standard for CLP Series Methods, VWR Chemicals BDH®
Supplier: VWR Chemicals BDH®
BDH0780-1ML
BDH0780-1MLEA 104.44 USD
BDH0780-1ML
Acid Surrogates Standard for CLP Series Methods, VWR Chemicals BDH®
Standards Environmental Analysis Standards
Semivolatiles standard.
CLP Series Methods: The US EPA retains analytical services through the Contract Laboratory Program (CLP). The CLP follows detailed SOPs derived from EPA methods and SW-846. The CLP Volatile Organic Analysis Method is an analytical method for the analysis of aqueous or solid samples for VOCs by GC/MS.