Alternate BTEX Standard for US EPA Method 602, VWR Chemicals BDH®
Supplier: VWR Chemicals BDH®
BDH0776-1ML
BDH0776-1MLEA 48.75 USD
BDH0776-1ML
Alternate BTEX Standard for US EPA Method 602, VWR Chemicals BDH®
Standards Environmental Analysis Standards
Volatiles standard.
US EPA Method 602 is a general analytical method for the monitoring of halogenated aromatic compounds in municipal and industrial wastewater by GC/PID.