BTEX Standard for US EPA Method 602
Supplier: VWR Chemicals BDH®
BDH0774-1ML BDH0777-1ML
BDH0774-1MLEA 28.88 USD
BDH0774-1ML BDH0777-1ML
BTEX Standard for US EPA Method 602
Standards Environmental Analysis Standards
Volatiles standard.
US EPA Method 602 is a general analytical method for the monitoring of halogenated aromatic compounds in municipal and industrial wastewater by GC/PID.