Polynuclear Aromatic Hydrocarbon (PAH) Combination Mix for US EPA Method 8310
Supplier: VWR Chemicals BDH®
BDH0772-1ML
BDH0772-1MLEA 106.05 USD
BDH0772-1ML
Polynuclear Aromatic Hydrocarbon (PAH) Combination Mix for US EPA Method 8310
Standards Environmental Analysis Standards
Semivolatiles standard.
US EPA Method 8310 is an analytical method for the monitoring of PNA hydrocarbons in groundwater and waste samples by HPLC using UV and fluorescence detectors.