Polynuclear Aromatic Hydrocarbon (PAH) Mix for US EPA Method 8270
Supplier: VWR Chemicals BDH®
BDH0771-1ML
BDH0771-1MLEA 143.9 USD
BDH0771-1ML
Polynuclear Aromatic Hydrocarbon (PAH) Mix for US EPA Method 8270
Standards Environmental Analysis Standards
Semivolatiles standard.
US EPA Method 8270 is a general analytical method for the monitoring of semivolatiles in environmental and waste samples by GC/MS.