Surrogate Standard for US EPA Method 8260
Supplier: VWR Chemicals BDH®
BDH0768-1ML
BDH0768-1MLEA 41.78 USD
BDH0768-1ML
Surrogate Standard for US EPA Method 8260
Standards Environmental Analysis Standards
Volatiles standard.
US EPA Method 8260 is a general analytical method for the monitoring of VOCs by GC/MS.