Phthalates Esters Semivolatiles Mix
Supplier: VWR Chemicals BDH®
BDH0762-1ML
BDH0762-1MLEA 124.75 USD
BDH0762-1ML
Phthalates Esters Semivolatiles Mix
Standards Environmental Analysis Standards
Semivolatiles standard.
US EPA Method 8061 is an analytical method for the monitoring of phthalate esters in aqueous and solid samples by GC/MS or GC/ECD.