Alcohols Mix for US EPA Method 8015, VWR Chemicals BDH®
Supplier: VWR Chemicals BDH®
BDH0760-1ML
BDH0760-1MLEA 48.75 USD
BDH0760-1ML
Alcohols Mix for US EPA Method 8015, VWR Chemicals BDH®
Standards Environmental Analysis Standards
Volatiles standard.
US EPA Method 8015 is a general analytical method for the monitoring of certain non-halogenated VOCs and SVOCs by GC/FID.