Purgeable Gases Mix B for US EPA Method 624
Supplier: VWR Chemicals BDH®
BDH0756-1ML
BDH0756-1MLEA 59.76 USD
BDH0756-1ML
Purgeable Gases Mix B for US EPA Method 624
Standards Environmental Analysis Standards
Volatiles standard.
US EPA Method 624 is a general analytical method for the monitoring of VOCs in municipal and industrial wastewater by GC/MS.