CLPV Series Volatile Analyte Mix A (High Level) for US EPA 502.1, 502.2, 524, 624, 8260, and CLP Series Methods
Supplier: VWR Chemicals BDH®
BDH0751-1ML
BDH0751-1MLEA 36.75 USD
BDH0751-1ML
CLPV Series Volatile Analyte Mix A (High Level) for US EPA 502.1, 502.2, 524, 624, 8260, and CLP Series Methods
Standards Environmental Analysis Standards
Volatiles standard.
US EPA Method 502.1 is a general analytical method for the monitoring of halogenated, portable VOCs in drinking water and raw water sources by GC with a Hall Detector.
US EPA Method 502.2 is a general analytical method for the monitoring of halogenated, purgeable VOCs in drinking water and raw water sources by GC megabore capillary column techniques and Hall and Photoionization Detectors (PID).
US EPA Method 524 is a GC/MS method for the analysis of purgeable VOCs in finished drinking water. Analytes added to Methanol 524.3 Revision 4 are supplied in a 24-component mix.
US EPA Method 624 is a general analytical method for the monitoring of VOCs in municipal and industrial wastewater by GC/MS.
US EPA Method 8260 is a general analytical method for the monitoring of VOCs by GC/MS.
CLP Series Methods: The US EPA retains analytical services through the Contract Laboratory Program (CLP). The CLP follows detailed SOPs derived from EPA methods and SW-846. The CLP Volatile Organic Analysis Method is an analytical method for the analysis of aqueous or solid samples for VOCs by GC/MS.