Acids Matrix Spike for US EPA Methods 625, 8270 and CLP Series Methods, VWR Chemicals BDH®
Supplier: VWR Chemicals BDH®
BDH0740-1ML
BDH0740-1MLEA 93.45 USD
BDH0740-1ML
Acids Matrix Spike for US EPA Methods 625, 8270 and CLP Series Methods, VWR Chemicals BDH®
Standards Environmental Analysis Standards
Semivolatiles standard.
US EPA Method 625 is an analytical method for the monitoring of acid and base neutral extractable fractions in municipal and industrial wastewater by GC/MS.
US EPA Method 8270 is a general analytical method for the monitoring of semivolatiles in environmental and waste samples by GC/MS.
CLP Series Methods: The US EPA retains analytical services through the Contract Laboratory Program (CLP). The CLP follows detailed SOPs derived from EPA methods and SW-846. The CLP Volatile Organic Analysis Method is an analytical method for the analysis of aqueous or solid samples for VOCs by GC/MS.