Surrogate Standard for US EPA 525
Supplier: VWR Chemicals BDH®
BDH0731-1ML
BDH0731-1MLEA 51.06 USD
BDH0731-1ML
Surrogate Standard for US EPA 525
Standards Environmental Analysis Standards
Semivolatiles standard.
US EPA Method 525 is an analytical method for the monitoring of semivolatiles in drinking water and raw water sources by GC/MS.